Merck
  • A t(4;6)(q12;p23) translocation disrupts a membrane-associated O-acetyl transferase gene (MBOAT1) in a patient with a novel brachydactyly-syndactyly syndrome.
A t(4;6)(q12;p23) translocation disrupts a membrane-associated O-acetyl transferase gene (MBOAT1) in a patient with a novel brachydactyly-syndactyly syndrome.
European journal of human genetics : EJHG (2007-04-19)
Johannes G Dauwerse, Bert B A de Vries, Cokkie H Wouters, Egbert Bakker, Gudrun Rappold, Geert R Mortier, Martijn H Breuning, Dorien J M Peters
ZUSAMMENFASSUNG
Here, we report a patient with a novel brachydactyly-syndactyly syndrome and a de novo translocation 46,XY,t(4;6)(q12;p23). We mapped the breakpoint and identified genes in the breakpoint region. One of the genes on chromosome 6, the membrane-associated O-acetyl transferase gene 1 (MBOAT1), was disrupted by the breakpoint. This gene consists of 13 exons and encodes a protein of 495 amino acids. MBOAT1 is predicted to be a transmembrane protein and belongs to the superfamily of membrane-bound O-acyltransferases. These proteins transfer organic compounds, usually fatty acids, onto hydroxyl groups of membrane-embedded targets. Identification of the transferred acyl group and the target may reveal the signaling pathways altered in this novel brachydactyly-syndactyly syndrome.