Merck
  • Enzymatic resolution of new carbonate intermediates for the synthesis of (< i> S</i>)-(+)-zopiclone.
Enzymatic resolution of new carbonate intermediates for the synthesis of (&lt; i&gt; S&lt;/i&gt;)-(+)-zopiclone.
Tetrahedron Asymmetry (2002)
Solares LF, et al.