- Design and synthesis of rhodamine 110 derivative and caspase-3 substrate for enzyme and cell-based fluorescent assay.
Design and synthesis of rhodamine 110 derivative and caspase-3 substrate for enzyme and cell-based fluorescent assay.
Bioorganic & medicinal chemistry letters (2001-01-05)
S X Cai, H Z Zhang, J Guastella, J Drewe, W Yang, E Weber
PMID11140728
ZUSAMMENFASSUNG
N-Octyloxycarbonyl-R110 (1), with enhanced cell penetration and retention properties, was prepared from rhodamine 110. The tetrapeptide substrate N-Ac-DEVD-N'-octyloxycarbonyl-R110 (3) was prepared and shown to be efficiently cleaved by human recombinant caspase-3 and by apoptotic HL-60 cells. This substrate should prove useful in cell-based assays for apoptosis inducers and inhibitors.