Merck
  • Analysis of fluorescent nanostructures in biological systems by means of spectral position determination microscopy (SPDM).
Analysis of fluorescent nanostructures in biological systems by means of spectral position determination microscopy (SPDM).
Current Microscopy Contributions to Advances in Science and Technology (2012)
Muller, P., et al., Mendez-Vilas, A.