Merck
  • Reactions of trimethylsilyl isocyanate and isothiocyanate with 3-(dialkylamino)-2H-azirines. A facile synthesis of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones and 4-(dialkylamino) imidazoline-2-thiones
Reactions of trimethylsilyl isocyanate and isothiocyanate with 3-(dialkylamino)-2H-azirines. A facile synthesis of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones and 4-(dialkylamino) imidazoline-2-thiones
The Journal of Organic Chemistry (1988)
Handke I, et al.