Merck
  • Interaction Between Natural Organic Matter (NOM) and (PAH) Comparison of Fluorescence Quenching and SPME
Interaction Between Natural Organic Matter (NOM) and (PAH) Comparison of Fluorescence Quenching and SPME
Fresenius Journal of Analytical Chemistry (1999)
Doll TE, et al.