Merck
  • Kinetic enzymatic determination of glycerol in wine and beer using a sequential injection system with spectrophotometric detection.
Kinetic enzymatic determination of glycerol in wine and beer using a sequential injection system with spectrophotometric detection.
Journal of agricultural and food chemistry (2006-06-08)
Hugo M Oliveira, Marcela A Segundo, José L F C Lima, Viviane Grassi, Elias A G Zagatto
ZUSAMMENFASSUNG
A sequential injection system for the automatic determination of glycerol in wine and beer was developed. The method is based on the rate of formation of NADH from the reaction of glycerol and NAD+ catalyzed by the enzyme glycerol dehydrogenase in solution. The determination of glycerol was performed between 0.3 and 3.0 mmol L(-1) (0.028 and 0.276 g L(-1)), and good repeatability was attained (rsd < 3.6%, n = 5) for all samples tested. The determination rate was 54 h(-1), the reagent consumption was only 0.75 micromol of NAD+ and 5.4 ng of enzyme per assay, and the waste production was 2.12 mL per assay. Results obtained for samples were in agreement with those obtained with the batch enzymatic method.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin-Dehydrogenase aus Cellulomonas sp., lyophilized powder, ≥50 units/mg protein