Merck
  • Differential regulation of macropinocytosis in macrophages by cytokines: implications for foam cell formation and atherosclerosis.
Differential regulation of macropinocytosis in macrophages by cytokines: implications for foam cell formation and atherosclerosis.
Cytokine (2013-06-25)
Daryn R Michael, Tim G Ashlin, Charlotte S Davies, Hayley Gallagher, Thomas W Stoneman, Melanie L Buckley, Dipak P Ramji
ZUSAMMENFASSUNG
A key event during the formation of lipid-rich foam cells during the progression of atherosclerosis is the uptake of modified low-density lipoproteins (LDL) by macrophages in response to atherogenic mediators in the arterial intima. In addition to scavenger receptor-dependent uptake of LDL, macropinocytosis is known to facilitate the uptake of LDL through the constitutive and passive internalization of large quantities of extracellular solute. In this study we confirm the ability of macropinocytosis to facilitate the uptake of modified LDL by human macrophages and show its modulation by TGF-β, IFN-γ, IL-17A and IL-33. Furthermore we show that the TGF-β-mediated inhibition of macropinocytosis is a Smad-2/-3-independent process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glyceraldehyd-3-Phosphat-Dehydrogenase aus Kaninchenmuskel, lyophilized powder, ≥75 units/mg protein
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes, lyophilized powder, 50-150 units/mg protein