Merck
Safeguards for cell cooperation in mouse embryogenesis shown by genome-wide cheater screen.
Science (New York, N.Y.) (2013-09-14)
Marion Dejosez, Hiroki Ura, Vicky L Brandt, Thomas P Zwaka
ZUSAMMENFASSUNG
Ensuring cooperation among formerly autonomous cells has been a central challenge in the evolution of multicellular organisms. One solution is monoclonality, but this option still leaves room for exploitative behavior, as it does not eliminate genetic and epigenetic variability. We therefore hypothesized that embryonic development must be protected by robust regulatory mechanisms that prevent aberrant clones from superseding wild-type cells. Using a genome-wide screen in murine induced pluripotent stem cells, we identified a network of genes (centered on p53, topoisomerase 1, and olfactory receptors) whose down-regulation caused the cells to replace wild-type cells in vitro and in the mouse embryo--without perturbing normal development. These genes thus appear to fulfill an unexpected role in fostering cell cooperation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Topoisomerase I human, buffered aqueous glycerol solution, ≥2 units/μL