Merck
The VDR gene FokI polymorphism and ovarian cancer risk.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2013-10-01)
Hui Xu, Su Li, Jian-Qing Qiu, Xiao-Lin Gao, Ping Zhang, Yong-Xia Yang
ZUSAMMENFASSUNG
The polymorphism of vitamin D receptor (VDR) gene is demonstrated to affect the activity of its encoding protein and the subsequent downstream effects mediated by vitamin D. Mutations in VDR gene FokI have been suggested in the development of various cancers. Whether the polymorphism of the VDR gene FokI confers risk to ovarian cancer still remains controversial across the published studies in different ethnicity. The aim of this meta-analysis was to determine the role of VDR gene FokI variant in the susceptibility to ovarian cancer. Six publications with 14 individual case-control studies involving a total of 10,964 subjects were finally included into our study after a comprehensive literature search of the PubMed, Embase, Web of Science, and Wanfang databases. The strength of the association between the VDR gene FokI polymorphism and ovarian cancer risk was estimated under the allelic (T vs. C), homozygous (TT vs. CC), additive (CT vs. CC), recessive (TT vs. CC + CT), and dominant (CT + TT vs. CC) gene models. The overall odds ratios (ORs) for the contrast models of T vs. C, TT vs. CC, CT vs. CC, and CT + TT vs. CC indicated that the VDR gene FokI variant was related to an increased risk of ovarian cancer (OR(T vs. C) = 1.09, 95 % confidence interval (CI) 1.03-1.15, P(OR) = 0.004; OR(TT vs. CC) = 1.17, 95 % CI 1.04-1.32, P(OR) = 0.011; OR(CT vs. CC) = 1.10, 95 % CI 1.01-1.20, P(OR) = 0.027; OR(CT + TT vs. CC) = 1.12, 95 % CI 1.03-1.21, P(OR) = 0.007). The stratified analysis among the Caucasians also identified a significant association between the VDR gene FokI polymorphism and the susceptibility to ovarian cancer. The present meta-analysis with large available published data has revealed that the VDR gene FokI polymorphism confers susceptibility to ovarian cancer, particularly among the Caucasian population.