Merck
Large changes in NAD levels associated with CD38 expression during HL-60 cell differentiation.
Biochemical and biophysical research communications (2013-11-13)
Zainab N Al-Abady, Barbara Durante, A John Moody, Richard A Billington
ZUSAMMENFASSUNG
NAD is an important cofactor involved in multiple metabolic reactions and as a substrate for several NAD-dependent signalling enzymes. One such enzyme is CD38 which, alongside synthesising Ca(2+)-releasing second messengers and acting as a cell surface receptor, has also been suggested to play a key role in NAD(+) homeostasis. CD38 is well known as a negative prognostic marker in B-CLL but the role of its enzymatic activity has not been studied in depth to date. We have exploited the HL-60 cell line as a model of inducible CD38 expression, to investigate CD38-mediated regulation intracellular NAD(+) levels and the consequences of changes in NAD(+) levels on cell physiology. Intracellular NAD(+) levels fell with increasing CD38 expression and this was reversed with the CD38 inhibitor, kuromanin confirming the key role of CD38 in NAD(+) homeostasis. We also measured the consequences of CD38 expression during the differentiation on a number of functions linked to NAD(+) and we show that some but not all NAD(+)-dependent processes are significantly affected by the lowered NAD(+) levels. These data suggest that both functional roles of CD38 might be important in the pathogenesis of B-CLL.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
NADase from porcine brain, ≥0.007 unit/mg solid