- [Conservation and divergence of cytoplasmic splicing in the organisms].
[Conservation and divergence of cytoplasmic splicing in the organisms].
Seikagaku. The Journal of Japanese Biochemical Society (2014-02-11)
Nozomu Koizumi, Yukihiro Nagashima
PMID24505943
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lysozym aus Hühnereiweiss, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme human, Lysobac™, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozym aus human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)