Merck
The human peptidylarginine deiminases type 2 and type 4 have distinct substrate specificities.
Biochimica et biophysica acta (2014-03-07)
Constance Assohou-Luty, Reinout Raijmakers, Willemien E Benckhuijsen, Judith Stammen-Vogelzangs, Arnoud de Ru, Peter A van Veelen, Kees L M C Franken, Jan W Drijfhout, Ger J M Pruijn
ZUSAMMENFASSUNG
Human peptidylarginine deiminases (hPADs) have been implicated in several diseases, particularly in rheumatoid arthritis. Since hPAD2 and hPAD4 are the isotypes expressed in the inflamed joints of RA patients and protein citrullination by PADs has been proposed to play a pathophysiological role, they represent unique therapeutic targets. To facilitate the development of substrate-based PAD inhibitors the substrate specificity of hPAD2 and hPAD4 was determined. Recombinant hPADs were expressed in bacteria or mammalian cell lines and allowed to citrullinate proteins in cell lysates, as well as a series of synthetic peptides. The citrullinated residues in proteins and the efficiency of peptide citrullination were determined by mass spectrometry. In total 320 hPAD2 and 178 hPAD4 citrullination sites were characterized. Amino acid residues most commonly found in citrullination sites for both isotypes are Gly at +1 and Tyr at +3 relative to the target arginine. For hPAD4 several additional amino acids were observed to be preferred at various positions from -4 to +4. The substrate motifs determined by amino acid substitution analysis partially confirmed these preferences, although peptide context dependent differences were also observed. Taken together, our data show that the enzyme specificity for cellular substrates and synthetic peptides differs for hPAD2 and hPAD4. hPAD4 shows more restrictive substrate specificity compared to hPAD2. Consensus sequences, which can be used as the basis for the development of PAD inhibitors, were derived for the citrullination sites of both hPAD2 and hPAD4.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Arginin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, reagent grade, ≥98%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Tyrosin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Glycin -hydrochlorid, ≥99% (HPLC)
Sigma-Aldrich
L-Arginin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Citrullin, ≥98% (TLC)
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Arginin, 99%, FCC, FG
SAFC
L-Arginin
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Supelco
L-Arginin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Tyrosin
Sigma-Aldrich
Glycin, 99%, FCC
Supelco
L-Tyrosin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
USP
L-Tyrosin, United States Pharmacopeia (USP) Reference Standard
Arginin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycin 1 M -Lösung
Sigma-Aldrich
Peptidyl-Arginin-Deiminase aus rabbit skeletal muscle, buffered aqueous glycerol solution, ≥200 units/mg protein (Bradford)
Sigma-Aldrich
L-Tyrosin, FG