Merck
  • Spacing and site isolation of amine groups in 3-aminopropyl-grafted silica materials: The role of protecting groups.
Spacing and site isolation of amine groups in 3-aminopropyl-grafted silica materials: The role of protecting groups.
Chemistry of Materials (2006)
Hicks JC, et al.