- Design, synthesis and biological evaluation of indane-2-arylhydrazinylmethylene-1,3-diones and indol-2-aryldiazenylmethylene-3-ones as beta-amyloid aggregation inhibitors.
Design, synthesis and biological evaluation of indane-2-arylhydrazinylmethylene-1,3-diones and indol-2-aryldiazenylmethylene-3-ones as beta-amyloid aggregation inhibitors.
European journal of medicinal chemistry (2010-02-09)
Marco Catto, Rosaria Aliano, Angelo Carotti, Saverio Cellamare, Fausta Palluotto, Rosa Purgatorio, Angelo De Stradis, Francesco Campagna
PMID20137834
ZUSAMMENFASSUNG
Biological screening of (hetero)aromatic compounds allowed the identification of some novel inhibitors of Abeta(1-40) aggregation, bearing indane and indole rings as common scaffolds. Molecular decoration of lead compounds led to inhibitors exhibiting a potency, measured by the Thioflavin T fluorimetric assay, ranging from high to low micromolar IC(50). The 2-(p-isopropylphenyldiazenylmethylene)indolone derivative 6c resulted as the most potent aggregation inhibitor exhibiting an IC(50) of 1.4 muM, with complete lack of fibril formation as confirmed by transmission electron microscopy. Structure-activity relationships suggested that binding to the Abeta peptide may be largely guided by pi-stacking and hydrogen bond interactions.