Merck
  • Method for determination of uranium isotopes in environmental samples by liquid-liquid extraction with triisooctylamine/xylene in hydrochloric media and alpha spectrometry.
Method for determination of uranium isotopes in environmental samples by liquid-liquid extraction with triisooctylamine/xylene in hydrochloric media and alpha spectrometry.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2012-08-09)
L Popov
ZUSAMMENFASSUNG
Alternative method for determination of uranium isotopes in various environmental samples is presented. The method is based on total decomposition of the solid materials and preconcentration of liquid samples. The separation of uranium from interfering radionuclides and stable matrix elements is attained by liquid-liquid extraction with triisooctylamine/xylene in hydrochloric media. After the additional removal of stable iron by extraction with diisopropyl ether, purified uranium is electrodeposited on stainless steel disks and measured by alpha spectrometry. The analytical method has been successfully applied to the determination of uranium isotopes in water and bottom sediments from the rivers Danube, Ogosta and Tzibritza in Northwestern Bulgaria. The analytical quality was checked by analyzing reference materials with different matrices.