Merck
Transfer-free growth of few-layer graphene by self-assembled monolayers.
Advanced materials (Deerfield Beach, Fla.) (2011-09-02)
Hyeon-Jin Shin, Won Mook Choi, Seon-Mi Yoon, Gang Hee Han, Yun Sung Woo, Eun Sung Kim, Seung Jin Chae, Xiang-Shu Li, Anass Benayad, Duong Dinh Loc, Fethullah Gunes, Young Hee Lee, Jae-Young Choi
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trichlor(octadecyl)silan, ≥90%
Sigma-Aldrich
Trimethoxyphenylsilan, 97%