Merck
  • Expression, crystallization and preliminary X-ray diffraction analysis of human paired Ig-like type 2 receptor alpha (PILRalpha).
Expression, crystallization and preliminary X-ray diffraction analysis of human paired Ig-like type 2 receptor alpha (PILRalpha).
Acta crystallographica. Section F, Structural biology and crystallization communications (2007-12-22)
Shigekazu Tabata, Kimiko Kuroki, Nobuo Maita, Jing Wang, Ikuo Shiratori, Hisashi Arase, Daisuke Kohda, Katsumi Maenaka
ZUSAMMENFASSUNG
Human paired immunoglobulin-like (Ig-like) type 2 receptor alpha (PILRalpha) is a type I membrane protein that is mainly expressed in immune-related cells such as monocytes, granulocytes and dendritic cells. PILRalpha can suppress the functions of such immune cells because it has the immunoreceptor tyrosine-based inhibitory motif (ITIM) in the intracellular region, which recruits the phosphatase Src homology-2 (SH2) domain-containing protein tyrosine phosphatase 2 (SHP-2) to inhibit phosphorylations induced by activation signals. The extracellular region of human PILRalpha comprises one immunoglobulin superfamily V-set domain and a stalk region. The V-set domain (residues 13-131) of human PILRalpha was overexpressed in Escherichia coli as inclusion bodies, refolded by rapid dilution and purified. The PILRalpha protein was successfully crystallized at 293 K using the sitting-drop vapour-diffusion method. The crystals diffracted to 1.3 A resolution at SPring-8 BL41XU; they belong to space group P2(1)2(1)2(1), with unit-cell parameters a = 40.4, b = 45.0, c = 56.9 A, and contain one molecule per asymmetric unit.