Merck
PILRalpha is a herpes simplex virus-1 entry coreceptor that associates with glycoprotein B.
Cell (2008-03-25)
Takeshi Satoh, Jun Arii, Tadahiro Suenaga, Jing Wang, Amane Kogure, Junji Uehori, Noriko Arase, Ikuo Shiratori, Shinya Tanaka, Yasushi Kawaguchi, Patricia G Spear, Lewis L Lanier, Hisashi Arase
ZUSAMMENFASSUNG
Glycoprotein B (gB) is one of the essential components for infection by herpes simplex virus-1 (HSV-1). Although several cellular receptors that associate with glycoprotein D (gD), such as herpes virus entry mediator (HVEM) and Nectin-1, have been identified, specific molecules that mediate HSV-1 infection by associating with gB have not been elucidated. Here, we found that paired immunoglobulin-like type 2 receptor (PILR) alpha associates with gB, and cells transduced with PILRalpha become susceptible to HSV-1 infection. Furthermore, HSV-1 infection of human primary cells expressing both HVEM and PILRalpha was blocked by either anti-PILRalpha or anti-HVEM antibody. Our results demonstrate that cellular receptors for both gB and gD are required for HSV-1 infection and that PILRalpha plays an important role in HSV-1 infection as a coreceptor that associates with gB. These findings uncover a crucial aspect of the mechanism underlying HSV-1 infection.