Merck
  • The effects of simultaneous treatment of SiO2 gate and Au electrode with octadecyltrichlorosilane and charge transfer molecules on characteristics of pentacene thin film transistors.
The effects of simultaneous treatment of SiO<SUB>2</SUB> gate and Au electrode with octadecyltrichlorosilane and charge transfer molecules on characteristics of pentacene thin film transistors.
Thin Solid Films (2005)
Pyo KS and Song CK.