Merck
  • Stereochemically defined C-substituted glutamic acids and their derivatives. 1. An efficient asymmetric synthesis of (2S, 3S)-3-methyl-and-3-trifluoromethylpyroglutamic acids.
Stereochemically defined <I>C-</I>substituted glutamic acids and their derivatives. 1. An efficient asymmetric synthesis of (2<I>S</I>, 3<I>S</I>)-3-methyl-and-3-trifluoromethylpyroglutamic acids.
Tetrahedron (1999)
Soloshonok VA, et al.