Merck
  • Enhanced carbon dioxide capture upon incorporation of N, N?-dimethylethylenediamine in the metal?organic framework CuBTTri.
Enhanced carbon dioxide capture upon incorporation of N, N?-dimethylethylenediamine in the metal?organic framework CuBTTri.
Chemical Science (2011)
McDonald TM, et al.