Merck
  • An optimal extraction solvent and purification adsorbent to produce anthocyanins from black rice (Oryza sativa cv. Heugjinjubyeo).
An optimal extraction solvent and purification adsorbent to produce anthocyanins from black rice (Oryza sativa cv. Heugjinjubyeo).
Food Technology and Biotechnology (2014)
Kang, You Jin, Seung Won Jung, and Seung Ju Lee.