Merck
  • Aqueous Dispersion and Temperature Induced Reversible Fluorescence Properties of 1-Pyrenecarboxaldehyde
Aqueous Dispersion and Temperature Induced Reversible Fluorescence Properties of 1-Pyrenecarboxaldehyde
International Letters of Chemistry, Physics and Astronomy (2013)
Ashaduzzaman, Md, and Masashi Kunitake