Merck
Ethylene formation from methional.
Biochemical and biophysical research communications (1978-03-30)
W A Pryor, R H Tang
PMID666768
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(Methylthio)propionaldehyd, 96%