Merck
  • Adsorption of supercritical carbon dioxide+ 2, 6-and 2, 7-dimethylnaphthalene isomers on NaY-type zeolite.
Adsorption of supercritical carbon dioxide+ 2, 6-and 2, 7-dimethylnaphthalene isomers on NaY-type zeolite.
Industrial & Engineering Chemistry Research (2003)
Iwai Y, et al.