Merck
  • Cardiac MRI assessment of atrial fibrosis in atrial fibrillation: implications for diagnosis and therapy.
Cardiac MRI assessment of atrial fibrosis in atrial fibrillation: implications for diagnosis and therapy.
Heart (British Cardiac Society) (2013-04-27)
Koji Higuchi, Mehmet Akkaya, Nazem Akoum, Nassir F Marrouche
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis