Merck
Sensitization of Hep3B hepatoma cells to cisplatin and doxorubicin by corilagin.
Phytotherapy research : PTR (2013-08-06)
Roberto Gambari, Desmond Kwok-Po Hau, Wai-Yeung Wong, Chung-Hin Chui
ZUSAMMENFASSUNG
The anticancer action of gallotannins is a well-developed topic. We have demonstrated the in vivo antitumour activity of corilagin on Hep3B hepatoma using the xenograft athymic nude mice model. Here, we further report the potential sensitization of Hep3B hepatoma cells to cisplatin and doxorubicin by corilagin. Our results showed that corilagin is able to enhance the cytotoxicity of both cisplatin and doxorubicin on the Hep3B hepatoma cells. We speculate the possible use of corilagin in combination with low dosages of the anticancer chemotherapeutic standard drugs like cisplatin and doxorubicin, with the aim of obtaining an increment in the anticancer effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Corilagin
Supelco
Corilagin, analytical standard