Merck
  • Type-1 hepatorenal syndrome associated with infections in cirrhosis: natural history, outcome of kidney function, and survival.
Type-1 hepatorenal syndrome associated with infections in cirrhosis: natural history, outcome of kidney function, and survival.
Hepatology (Baltimore, Md.) (2013-09-17)
Rogelio Barreto, Claudia Fagundes, Mónica Guevara, Elsa Solà, Gustavo Pereira, Ezequiel Rodríguez, Isabel Graupera, Marta Martín-Llahí, Xavier Ariza, Andrés Cárdenas, Javier Fernández, Juan Rodés, Vicente Arroyo, Pere Ginès
ZUSAMMENFASSUNG
Type-1 hepatorenal syndrome (HRS) is a common complication of bacterial infections in cirrhosis, but its natural history remains undefined. To assess the outcome of kidney function and survival of patients with type-1 HRS associated with infections, 70 patients diagnosed during a 6-year period were evaluated prospectively. Main outcomes were no reversibility of type-1 HRS during treatment of the infection and 3-month survival. Forty-seven (67%) of the 70 patients had no reversibility of type-1 HRS during treatment of the infection. [Correction to previous sentence added March 10, 2014, after first online publication: "Twenty-three (33%)" was changed to "Forty-seven (67%)."] The main predictive factor of no reversibility of type-1 HRS was absence of infection resolution (no reversibility: 96% versus 48% in patients without and with resolution of the infection; P < 0.001). Independent predictive factors of no reversibility of type-1 HRS were age, high baseline serum bilirubin, nosocomial infection, and reduction in serum creatinine <0.3 mg/dL at day 3 of antibiotic treatment. No reversibility was also associated with severity of circulatory dysfunction, as indicated by more marked activity of the vasoconstrictor systems. In the whole series, 3-month probability of survival was only 21%. Factors associated with poor prognosis were baseline serum bilirubin, no reversibility of type-1 HRS, lack of resolution of the infection, and development of septic shock after diagnosis of type-1 HRS. Type-1 HRS associated with infections is not reversible in two-thirds of patients with treatment of infection only. No reversibility of type-1 HRS is associated with lack of resolution of the infection, age, high bilirubin, and no early improvement of kidney function and implies a poor prognosis. These results may help advance the management of patients with type-1 HRS associated with infections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)