Merck
Long-term outcome of a metal-on-polyethylene cementless hip resurfacing.
The Journal of arthroplasty (2013-10-05)
Timothy L Tan, Edward Ebramzadeh, Patricia A Campbell, Mariam Al-Hamad, Harlan C Amstutz
ZUSAMMENFASSUNG
Due to the well-documented problems surrounding metal-on-metal bearings, the use of hip resurfacing has declined. Since the potential benefits of hip resurfacing remain desirable, it may be beneficial to investigate the long-term outcome of hip resurfacings using metal-on-polyethylene in the 1980's. We report the long-term survivorship and modes of failure of a cementless metal-on-polyethylene resurfacing (n = 178) with different porous ingrowth surfaces. While acetabular loosening was absent, a high incidence of femoral failures (femoral loosening = 18.1%, osteolytic neck fracture = 21%) occurred despite using the same ingrowth surface for both components. Ongoing developments using the lessons learned from these previous generation components and utilizing modern low wear materials, e.g., cross-linked polyethylene, may lead to improved implants for future hip resurfacings.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylen, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylen, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylen, High density, melt index 12 g/10 min (190 °C/2.16kg)
Polyethylen (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylen, Medium density
Sigma-Aldrich
Polyethylen, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylen, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Supelco
Polyethylen, analytical standard, suitable for gel permeation chromatography (GPC), 2,000