Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium Zubereitung, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium (in Sure/Seal™ Flasche), in a Sure/Seal™ bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Sigma-Aldrich
Magnesium (in Sure/Seal™ Flasche), in a Sure/Seal™ bottle, turnings, 37.5 mmol
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 1m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%