Merck
Epinephrine for cardiac arrest: are we doing more harm than good?
Anesthesiology (2013-10-19)
Vijay Krishnamoorthy, Monica S Vavilala, Michael R Fettiplace, Guy Weinberg
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Epinephrin
Adrenalin, European Pharmacopoeia (EP) Reference Standard