Merck
  • Efficient extra- and intracellular alkalinization improves cardiovascular functions in severe lactic acidosis induced by hemorrhagic shock.
Efficient extra- and intracellular alkalinization improves cardiovascular functions in severe lactic acidosis induced by hemorrhagic shock.
Anesthesiology (2013-11-23)
Antoine Kimmoun, Nicolas Ducrocq, Nacira Sennoun, Khodr Issa, Charlène Strub, Jean-Marie Escanyé, Sébastien Leclerc, Bruno Levy
ZUSAMMENFASSUNG
Lactic acidosis is associated with cardiovascular failure. Buffering with sodium bicarbonate is proposed in severe lactic acidosis. Bicarbonate induces carbon dioxide generation and hypocalcemia, both cardiovascular depressant factors. The authors thus investigated the cardiovascular and metabolic effects of an adapted sodium bicarbonate therapy, including prevention of carbon dioxide increase with hyperventilation and ionized calcium decrease with calcium administration. Lactic acidosis was induced by hemorrhagic shock. Twenty animals were randomized into five groups: (1) standard resuscitation with blood retransfusion and norepinephrine (2) adapted sodium bicarbonate therapy (3) nonadapted sodium bicarbonate therapy (4) standard resuscitation plus calcium administration (5) hyperventilation. Evaluation was focused in vivo on extracellular pH, on intracellular pH estimated by P nuclear magnetic resonance and on myocardial contractility by conductance catheter. Aortic rings and mesenteric arteries were isolated and mounted in a myograph, after which arterial contractility was measured. All animals in the hyperventilation group died prematurely and were not included in the statistical analysis. When compared with sham rats, shock induced extracellular (median, 7.13; interquartile range, [0.10] vs. 7.30 [0.01]; P = 0.0007) and intracellular acidosis (7.26 [0.18] vs. 7.05 [0.13]; P = 0.0001), hyperlactatemia (7.30 [0.01] vs. 7.13 [0.10]; P = 0.0008), depressed myocardial elastance (2.87 [1.31] vs. 0.5 [0.53] mmHg/μl; P = 0.0001), and vascular hyporesponsiveness to vasoconstrictors. Compared with nonadapted therapy, adapted bicarbonate therapy normalized extracellular pH (7.03 [0.12] vs. 7.36 [0.04]; P < 0.05), increased intracellular pH to supraphysiological values, improved myocardial elastance (1.68 [0.41] vs. 0.72 [0.44] mmHg/μl; P < 0.05), and improved aortic and mesenteric vasoreactivity. A therapeutic strategy based on alkalinization with sodium bicarbonate along with hyperventilation and calcium administration increases pH and improves cardiovascular function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumbicarbonat -Lösung, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
(−)-Norepinephrin, ≥98%, crystalline
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
(±)-Norepinephrin -(+)-bitartratsalz
Sigma-Aldrich
DL-Norepinephrin -hydrochlorid, crystalline, ≥97% (TLC)
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Supelco
Natriumbicarbonat-Konzentrat, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
L-Norepinephrin -hydrochlorid, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C