Merck
  • The role of NMDA and mGluR5 receptors in calcium mobilization and neurotoxicity of homocysteine in trigeminal and cortical neurons and glial cells.
The role of NMDA and mGluR5 receptors in calcium mobilization and neurotoxicity of homocysteine in trigeminal and cortical neurons and glial cells.
Journal of neurochemistry (2013-11-26)
Polina A Abushik, Minna Niittykoski, Raisa Giniatullina, Anastasia Shakirzyanova, Genevieve Bart, Dmitriy Fayuk, Dmitry A Sibarov, Sergei M Antonov, Rashid Giniatullin
ZUSAMMENFASSUNG
Recent studies suggested contribution of homocysteine (HCY) to neurodegenerative disorders and migraine. However, HCY effect in the nociceptive system is essentially unknown. To explore the mechanism of HCY action, we studied short- and long-term effects of this amino acid on rat peripheral and central neurons. HCY induced intracellular Ca²⁺ transients in cultured trigeminal neurons and satellite glial cells (SGC), which were blocked by the NMDA antagonist AP-5 in neurons, but not in SGCs. In contrast, 3-((2-Methyl-4-thiazolyl)ethynyl)pyridine (MTEP), the metabotropic mGluR5 (metabotropic glutamate receptor 5 subtype) antagonist, preferentially inhibited Ca²⁺ transients in SGCs. Prolonged application of HCY induced apoptotic cell death of both kinds of trigeminal cells. The apoptosis was blocked by AP-5 or by the mGluR5 antagonist MTEP. Likewise, in cortical neurons, HCY-induced cell death was inhibited by AP-5 or MTEP. Imaging with 2',7'-dichlorodihydrofluorescein diacetate or mitochondrial dye Rhodamine-123 as well as thiobarbituric acid reactive substances assay did not reveal involvement of oxidative stress in the action of HCY. Thus, elevation of intracellular Ca²⁺ by HCY in neurons is mediated by NMDA and mGluR5 receptors while SGC are activated through the mGluR5 subtype. Long-term neurotoxic effects in peripheral and central neurons involved both receptor types. Our data suggest glutamatergic mechanisms of HCY-induced sensitization and apoptosis of trigeminal nociceptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Homocystein, ≥98.0% (NT)