Merck
  • Hepatocellular carcinoma enhancement on contrast-enhanced CT and MR imaging: response assessment after treatment with sorafenib: preliminary results.
Hepatocellular carcinoma enhancement on contrast-enhanced CT and MR imaging: response assessment after treatment with sorafenib: preliminary results.
La Radiologia medica (2013-12-04)
Giuseppe Salvaggio, Alessandro Furlan, Francesco Agnello, Giuseppe Cabibbo, Daniele Marin, Lydia Giannitrapani, Chiara Genco, Massimo Midiri, Roberto Lagalla, Giuseppe Brancatelli
ZUSAMMENFASSUNG
This study was undertaken to compare response evaluation criteria in solid tumours (RECIST) 1.1 and modified RECIST (mRECIST) in patients with unresectable hepatocellular carcinoma (HCC) on sorafenib, and to describe HCC enhancement changes before and after sorafenib treatment. Seventeen patients (12 men, 5 women; mean age 69 years; age range 58-79 years) were included. Tumour response was assessed according to RECIST and mRECIST. Two readers placed a region of interest (ROI) within each target lesion, on the portion showing enhancement during the arterial phase. The lesion attenuation values measured within the ROIs on computed tomography or the signal intensity measured on magnetic resonance imaging, during the unenhanced phase, hepatic arterial phase and venous phase were recorded. Changes in arterial and venous contrast enhancement before and after treatment were compared among the mRECIST groups using Mann-Whitney U test. Agreement between mRECIST and RECIST was good (Cohen's k coefficient, 0.791). Patients with partial response had a greater decrease in arterial enhancement (-79.8%) than did patients with stable disease (SD) (-24.8%; p = 0.011) or progressive disease (PD) (-32.9%; p = 0.034). No statistically significant difference in arterial enhancement variation was found among patients with SD and PD. No statistically significant difference in venous enhancement was found among the mRECIST groups. mRECIST showed a more favourable response compared to RECIST 1.1 in patients with unresectable HCC receiving sorafenib.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
N-Methyl-D-Glucamin, 99.0-100.5% (titration)
Sigma-Aldrich
Nicotinamid, ≥99.5% (HPLC)
Supelco
Niacinamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamid, ≥98% (HPLC), powder
USP
Niacinamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Niacinamid, meets USP testing specifications
Sigma-Aldrich
N-Methyl-D-Glucamin, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
Meglumin, 99.0-100.5% dry basis, meets USP testing specifications
Supelco
Nicotinamid (Vitamin B3) -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nicotinamid, ≥98.5% (HPLC)
Nicotinamid, European Pharmacopoeia (EP) Reference Standard
Supelco
Meglumin, Pharmaceutical Secondary Standard; Certified Reference Material
Meglumin, European Pharmacopoeia (EP) Reference Standard