Merck
  • A particular set of small non-coding RNAs is bound to the distinctive Argonaute protein of Trypanosoma cruzi: insights from RNA-interference deficient organisms.
A particular set of small non-coding RNAs is bound to the distinctive Argonaute protein of Trypanosoma cruzi: insights from RNA-interference deficient organisms.
Gene (2014-01-28)
Maria Rosa Garcia-Silva, Julia Sanguinetti, Florencia Cabrera-Cabrera, Oscar Franzén, Alfonso Cayota
ZUSAMMENFASSUNG
The study of small RNAs and Argonaute proteins in eukaryotes that are deficient in functional RNA interference could provide insights into novel functions of small RNAs. In this study we describe small non-coding RNAs bound to a distinctive Argonaute protein of Trypanosoma cruzi, TcPIWI-tryp. Co-immunoprecipitation of TcPIWI-tryp followed by deep sequencing of isolated RNA identified abundant small RNAs derived from rRNAs and tRNAs. The small RNA repertoire differed from that of the canonical Argonaute in organisms with functional RNA interference, which could indicate novel biological functions for TcPIWI-tryp in T. cruzi and other members of the trypanosomatid clade.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ribonukleinsäure, Transfer aus Backhefe (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonukleinsäure, Transfer aus E. coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
Ribonukleinsäure, Transfer aus Backhefe (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonukleinsäure, Transfer aus Backhefe (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonukleinsäure, Transfer aus Rinderleber, Type XI, lyophilized powder
Sigma-Aldrich
Ribonukleinsäure, Transfer aus Weizenkeimen, Type V, 15-19 units/mg solid