Merck
  • Occurrence of endocrine-disrupting phenols and estrogens in water and sediment of the Songhua river, northeastern China.
Occurrence of endocrine-disrupting phenols and estrogens in water and sediment of the Songhua river, northeastern China.
Archives of environmental contamination and toxicology (2014-01-29)
Zifeng Zhang, Nanqi Ren, Kurunthachalam Kannan, Jun Nan, Liyan Liu, Wanli Ma, Hong Qi, Yifan Li
ZUSAMMENFASSUNG
Concentrations of six phenolic endocrine-disrupting chemicals [4-tert-octylphenol (OP), 4-t-nonylphenol (4-t-NP), 4-n-nonylphenol (4-n-NP), nonylphenol mono- to di-ethoxylates (NP1EO, NP2EO), and bisphenol A (BPA)] and five estrogens [estrone (E1), β-estradiol (E2), estriol (E3) 17α-ethynylestradiol (EE2), and diethylstilbestrol (DES)] were determined in surface water and sediment samples collected from the Songhua River in northeast China. Concentrations of sum of five alkylphenols and alkylphenol ethoxylates (ΣOP, 4-n-NP, 4-t-NP, NP1EO, NP2EO) were 117-1,030 ng L(-1) (mean 296) in water samples and 25.5-386 ng g(-1) (mean 67.3 ng g(-1) dry weight (dw)) in sediments. Concentrations of BPA in water and sediments were 8.24-263 ng L(-1) (mean 52.0) and 1.60-17.3 ng g(-1) dw (mean 4.90 dw), respectively. Concentrations in water were 0.840-20.8 ng L(-1) (mean 5.03) for the sum of three natural steroidal estrogens (ΣE1, E2, E3) and below detection limit (BDL) at -1.38 ng L(-1) (average 0.200) for the sum of two synthetic estrogens (EE2, ΣDES). Among estrogens, only E1 was detected in all of the sediment samples in the range of 0.100-3.00 ng g(-1) dw. Concentrations of Σphenolic EDCs and Σestrogens in water and sediments and their correlations with total organic carbon indicated that these contaminants originate from similar sources, such as municipal wastewater. In situ [Formula: see text] values and sediment-water fugacity fraction were calculated for the target chemicals, and the results indicated that these chemicals were, in general, supersaturated in sediments relative to those in water.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
4,4′-Isopropylidendiphenol, ≥99%
Sigma-Aldrich
4,4′-Isopropylidendiphenol, 97%
Sigma-Aldrich
4-tert-Octylphenol, 97%
Supelco
4,4′-Isopropylidendiphenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
4-Nonylphenol, PESTANAL®, analytical standard
Supelco
4-tert-Octylphenol, analytical standard
Supelco
4-tert.-Octylphenol -Lösung, 1000 μg/mL in acetone, analytical standard