Merck
  • The estrogenic effects of apigenin, phloretin and myricetin based on uterotrophic assay in immature Wistar albino rats.
The estrogenic effects of apigenin, phloretin and myricetin based on uterotrophic assay in immature Wistar albino rats.
Toxicology letters (2014-02-04)
Nurhayat Barlas, Saadet Özer, Gözde Karabulut
ZUSAMMENFASSUNG
Chemicals that occur in vegetal food and known as phytoestrogens, because of their structures similarity to estrogen, have benefits on chronic diseases. Despite this, when they are taken at high amounts, they can cause harmful effects on endocrine system of human and animals. In this study, it has been intended to determine the estrogenic potencies of phytoestrogens apigenin, phloretin and myricetin whose affinities for estrogen receptors in vitro. The female rats divided into 17 groups, each containing six rats. There was a negative control group and there were positive control dose groups which contains ethinyl estradiol, ethinyl estradiol+tamoxifen and genistein. The other dose groups which were tested for estrogenic activity contains apigenin, myricetin and phloretin All chemicals have been given to Wistar immature female rats with oral gavage for 3 consecutive days. By using uterotrophic analysis, uterus wet and blotted weights, vaginal opening, uterus length of female rats has been recorded at the end of the experiment. For detect of cell response, luminal epithelium height, gland number and lactoferrin intensity in luminal epithelium of uterus were evaluated. Biochemical analysises in blood were performed. Relative uterus weights of rats in 100 mg/kg/day dose group of myricetin were statistically increased according to vehicle control and positive control groups. In dose groups of apigenin and phloretin it was found that there were cell responses in uterus. All treatment groups had a significant difference in the high intensity of lactoferrin and uterine gland count compared to oil control group. There was no difference between phloretin and apigenin treatment groups in uterine weight statictically. Uterine heights were increased in positive control groups and 100 mg/kg/day dose group of myricetin. Epithelial cell heights were increased in treatment groups except apigenin and phloretin dose groups. There was no difference between all treatment groups in vaginal opening values according to positive control.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Phloretin, ≥99%
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Supelco
Apigenin, analytical standard
Supelco
Myricetin, analytical standard