Merck
Sugars and risk of mortality in the NIH-AARP Diet and Health Study.
The American journal of clinical nutrition (2014-02-21)
Natasha Tasevska, Yikyung Park, Li Jiao, Albert Hollenbeck, Amy F Subar, Nancy Potischman
ZUSAMMENFASSUNG
Although previous studies have linked intake of sugars with incidence of cancer and other chronic diseases, its association with mortality remains unknown. We investigated the association of total sugars, added sugars, total fructose, added fructose, sucrose, and added sucrose with the risk of all-cause, cardiovascular disease, cancer, and other-cause mortality in the NIH-AARP Diet and Health Study. The participants (n = 353,751), aged 50-71 y, were followed for up to 13 y. Intake of individual sugars over the previous 12 mo was assessed at baseline by using a 124-item NIH Diet History Questionnaire. In fully adjusted models (fifth quartile compared with first quartile), all-cause mortality was positively associated with the intake of total sugars [HR (95% CI): 1.13 (1.06, 1.20); P-trend < 0.0001], total fructose [1.10 (1.04, 1.17); P-trend < 0.0001], and added fructose [1.07 (1.01, 1.13); P-trend = 0.005) in women and total fructose [1.06 (1.01, 1.10); P-trend = 0.002] in men. In men, a weak inverse association was found between other-cause mortality and dietary added sugars (P-trend = 0.04), sucrose (P-trend = 0.03), and added sucrose (P-trend = 0.006). Investigation of consumption of sugars by source showed that the positive association with mortality risk was confined only to sugars from beverages, whereas the inverse association was confined to sugars from solid foods. In this large prospective study, total fructose intake was weakly positively associated with all-cause mortality in both women and men, whereas added sugar, sucrose, and added sucrose intakes were inversely associated with other-cause mortality in men. In our analyses, intake of added sugars was not associated with an increased risk of mortality. The NIH-AARP Diet and Health Study was registered at clinicaltrials.gov as NCT00340015.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Millipore
Saccharose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20
Saccharose, European Pharmacopoeia (EP) Reference Standard