Merck
  • Multi-element distribution profile in Sprague-Dawley rats: effects of intratracheal instillation of Cr(VI) and Zn intervention.
Multi-element distribution profile in Sprague-Dawley rats: effects of intratracheal instillation of Cr(VI) and Zn intervention.
Toxicology letters (2014-02-25)
Yanshuang Song, Tianjing Wang, Ji Pu, Jian Guo, Zhangjian Chen, Yun Wang, Guang Jia
ZUSAMMENFASSUNG
Our previous epidemiological study revealed that co-exposure of [Cr(VI)] with small amount of heavy metals could induce peripheral blood element imbalance, but little was known about the contribution of Cr(VI) itself and the multi-element distribution profile in other target tissues. We explored element homeostasis in the blood, RBC, serum and lung after Cr(VI) exposure and Zn intervention. 60 Sprague-Dawley male rats received intratracheal instillation of Cr(VI) (0, 0.063, 0.630mgCr/kg) weekly and/or intragastric administration of zinc sulfate (0, 10mgZn/kg) daily for one month. Element contents and urinary 8-hydroxy-2'-deoxyguanosine (8-OHdG) concentrations were determined. Dose-response relationship was observed among rats exposed to Cr(VI). Ca, Mg, Mn in the blood, Fe, Mg, Se in the serum, and Mg and Zn in lung tissue decreased significantly, while Ca, Co, Cr, Mg, Mn, Se in RBC, and Ca, Co, Mo in the lung increased after Cr(VI) exposure. The alteration trends manifested differently, with RBC the most sensitive. Cr induced increase of urinary 8-OHdG, which decreased after Zn intervention. Zn intervention could help to restore element homeostasis after Cr(VI) exposure, especially for Ca, Fe, Mg and Se.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinksulfat Heptahydrat, ACS reagent, 99%
Sigma-Aldrich
Kaliumchromat, ACS reagent, ≥99.0%
Sigma-Aldrich
Zinksulfat Heptahydrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinksulfat Heptahydrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Zinksulfat Heptahydrat, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinksulfat -Lösung, 0.3 N
Sigma-Aldrich
Chrom, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Zinksulfat Heptahydrat, ≥99.95% trace metals basis
Sigma-Aldrich
8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)
Sigma-Aldrich
Zinksulfat -Lösung, BioUltra, Molecular Biology, 2.0 M in H2O
Sigma-Aldrich
Chrom, powder, 99.5%, −100 mesh
Sigma-Aldrich
Zinksulfat Heptahydrat, suitable for plant cell culture
Sigma-Aldrich
Chrom, chips, 99.995% trace metals basis
Sigma-Aldrich
Zinksulfat Heptahydrat, meets USP testing specifications
Sigma-Aldrich
Kaliumchromat, ≥99.95% trace metals basis
Sigma-Aldrich
Chrom, chips, thickness ~2 mm, 99.5%
Chrom, rod, 25mm, diameter 2.0mm, 99.7+%
Chrom, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chrom, rod, 50mm, diameter 2.0mm, 99.7+%
Chrom, rod, 10mm, diameter 2.0mm, 99.7+%
Chrom, bar, 100mm x 2mm x 2mm, 99.7+%
Chrom, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chrom, microfoil, disks, 10mm, thinness 0.025μm, specific density 18.21μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chrom, rod, 25mm, diameter 5.0mm, 99.7+%
Chrom, foil, 100x100mm, thickness 3.0mm, hard, 99.95%
Chrom, microfoil, disks, 25mm, thinness 0.25μm, specific density 178.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chrom, microfoil, disks, 25mm, thinness 1.0μm, specific density 719μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chrom, rod, 10mm, diameter 5.0mm, 99.7+%
Chrom, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Chrom, rod, 100mm, diameter 2.0mm, 99.7+%