Merck
  • Acute toxicity of multi-walled carbon nanotubes, sodium pentachlorophenate, and their complex on earthworm Eisenia fetida.
Acute toxicity of multi-walled carbon nanotubes, sodium pentachlorophenate, and their complex on earthworm Eisenia fetida.
Ecotoxicology and environmental safety (2014-02-25)
Liujun Zhang, Changwei Hu, Weili Wang, Funian Ji, Yibin Cui, Mei Li
ZUSAMMENFASSUNG
Laboratory experiments were undertaken to relate biomarker responses to the toxicities of multi-walled carbon nanotubes (MWCNTs) and sodium pentachlorophenate (PCP-Na), both individually and combined. The acute toxicities of MWCNTs and PCP-Na on earthworm Eisenia fetida were studied through different exposure methods (filter paper contact test, immersion contact test, and artificial soil contact test). Enzyme activity and malondialdehyde (MDA) content in the earthworm E. fetida exposed to MWCNTs and PCP-Na in filter paper contact test, both individually and under combined exposure, were determined. After exposure, PCP-Na induced observable acute toxicity while the MWCNTs induced slight toxicity. Interestingly the earthworms exposed to the mixture of MWCNTs and PCP-Na demonstrated different expression of enzymatic biomarkers from those exposed to MWCNTs or PCP-Na alone. Our results indicated that the toxicity of PCP-Na on E. fetida may be alleviated by the appearance of MWCNTs for all exposure methods except for immersion contact test.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pentachlorphenol, 97%
Supelco
Pentachlorphenol -Lösung, certified reference material, 5000 μg/mL in methanol
Supelco
Pentachlorphenol, analytical standard