Merck
  • Safety of the combined use of praziquantel and albendazole in the treatment of human hydatid disease.
Safety of the combined use of praziquantel and albendazole in the treatment of human hydatid disease.
The American journal of tropical medicine and hygiene (2014-03-13)
Lucía Alvela-Suárez, Virginia Velasco-Tirado, Moncef Belhassen-Garcia, Ignacio Novo-Veleiro, Javier Pardo-Lledías, Angela Romero-Alegría, Luis Pérez del Villar, María Paz Valverde-Merino, Miguel Cordero-Sánchez
ZUSAMMENFASSUNG
There is still no well-established consensus about the clinical management of hydatidosis. Currently, surgery continues to be the first therapeutic option, although treatment with anti-parasitic drugs is indicated as an adjuvant to surgery to decrease the number of relapses and hydatid cyst size. When surgery is not possible, medical treatment is indicated. Traditionally, albendazole was used in monotherapy as the standard treatment. However, combined therapy with albendazole plus praziquantel appears to improve anti-parasitic effectiveness. To date, no safety studies focusing on such combined therapy have been published for the treatment of hydatidosis. In this work, we analyze the adverse effects seen in 57 patients diagnosed with hydatidosis who were treated with praziquantel plus albendazole combined therapy between 2006 and 2010.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Praziquantel, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Albendazol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Albendazol, analytical standard, ≥98%
Supelco
Praziquantel, VETRANAL®, analytical standard
USP
Albendazol, United States Pharmacopeia (USP) Reference Standard
Praziquantel, European Pharmacopoeia (EP) Reference Standard
Albendazol, European Pharmacopoeia (EP) Reference Standard
Praziquantel für die Systemeignung, European Pharmacopoeia (EP) Reference Standard