Merck
A novel prodrug-based strategy to increase effects of bumetanide in epilepsy.
Annals of neurology (2014-03-13)
Kathrin Töllner, Claudia Brandt, Manuel Töpfer, Gerda Brunhofer, Thomas Erker, Mario Gabriel, Peter W Feit, Jenna Lindfors, Kai Kaila, Wolfgang Löscher
ZUSAMMENFASSUNG
There is considerable interest in using bumetanide, a chloride importer Na-K-Cl cotransporter antagonist, for treatment of neurological diseases, such as epilepsy or ischemic and traumatic brain injury, that may involve deranged cellular chloride homeostasis. However, bumetanide is heavily bound to plasma proteins (~98%) and highly ionized at physiological pH, so that it only poorly penetrates into the brain, and chronic treatment with bumetanide is compromised by its potent diuretic effect. To overcome these problems, we designed lipophilic and uncharged prodrugs of bumetanide that should penetrate the blood-brain barrier more easily than the parent drug and are converted into bumetanide in the brain. The feasibility of this strategy was evaluated in mice and rats. Analysis of bumetanide levels in plasma and brain showed that administration of 2 ester prodrugs of bumetanide, the pivaloyloxymethyl (BUM1) and N,N-dimethylaminoethylester (BUM5), resulted in significantly higher brain levels of bumetanide than administration of the parent drug. BUM5, but not BUM1, was less diuretic than bumetanide, so that BUM5 was further evaluated in chronic models of epilepsy in mice and rats. In the pilocarpine model in mice, BUM5, but not bumetanide, counteracted the alteration in seizure threshold during the latent period. In the kindling model in rats, BUM5 was more efficacious than bumetanide in potentiating the anticonvulsant effect of phenobarbital. Our data demonstrate that the goal of designing bumetanide prodrugs that specifically target the brain is feasible and that such drugs may resolve the problems associated with using bumetanide for treatment of neurological disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pentylentetrazol
Sigma-Aldrich
Bumetanid, ≥98%
Bumetanid, European Pharmacopoeia (EP) Reference Standard