Merck
  • Striatal response to reward anticipation: evidence for a systems-level intermediate phenotype for schizophrenia.
Striatal response to reward anticipation: evidence for a systems-level intermediate phenotype for schizophrenia.
JAMA psychiatry (2014-03-14)
Oliver Grimm, Andreas Heinz, Henrik Walter, Peter Kirsch, Susanne Erk, Leila Haddad, Michael M Plichta, Nina Romanczuk-Seiferth, Lydia Pöhland, Sebastian Mohnke, Thomas W Mühleisen, Manuel Mattheisen, Stephanie H Witt, Axel Schäfer, Sven Cichon, Markus Nöthen, Marcella Rietschel, Heike Tost, Andreas Meyer-Lindenberg
ZUSAMMENFASSUNG
Attenuated ventral striatal response during reward anticipation is a core feature of schizophrenia that is seen in prodromal, drug-naive, and chronic schizophrenic patients. Schizophrenia is highly heritable, raising the possibility that this phenotype is related to the genetic risk for the disorder. To examine a large sample of healthy first-degree relatives of schizophrenic patients and compare their neural responses to reward anticipation with those of carefully matched controls without a family psychiatric history. To further support the utility of this phenotype, we studied its test-retest reliability, its potential brain structural contributions, and the effects of a protective missense variant in neuregulin 1 (NRG1) linked to schizophrenia by meta-analysis (ie, rs10503929). Examination of a well-established monetary reward anticipation paradigm during functional magnetic resonance imaging at a university hospital; voxel-based morphometry; test-retest reliability analysis of striatal activations in an independent sample of 25 healthy participants scanned twice with the same task; and imaging genetics analysis of the control group. A total of 54 healthy first-degree relatives of schizophrenic patients and 80 controls matched for demographic, psychological, clinical, and task performance characteristics were studied. Blood oxygen level-dependent response during reward anticipation, analysis of intraclass correlations of functional contrasts, and associations between striatal gray matter volume and NRG1 genotype. Compared with controls, healthy first-degree relatives showed a highly significant decrease in ventral striatal activation during reward anticipation (familywise error-corrected P < .03 for multiple comparisons across the whole brain). Supplemental analyses confirmed that the identified systems-level functional phenotype is reliable (with intraclass correlation coefficients of 0.59-0.73), independent of local gray matter volume (with no corresponding group differences and no correlation to function, and with all uncorrected P values >.05), and affected by the NRG1 genotype (higher striatal responses in controls with the protective rs10503929 C allele; familywise error-corrected P < .03 for ventral striatal response). Healthy first-degree relatives of schizophrenic patients show altered striatal activation during reward anticipation in a directionality and localization consistent with prior patient findings. This provides evidence for a functional neural system mechanism related to familial risk. The phenotype can be assessed reliably, is independent of alterations in striatal structure, and is influenced by a schizophrenia candidate gene variant in NRG1. These data encourage us to further investigate the genetic and molecular contributions to this phenotype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dopamin -hydrochlorid
Supelco
Dopamin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dopamin -hydrochlorid -Lösung, 1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Dopamin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard