Merck
  • Reproducibility of estradiol and testosterone levels in postmenopausal women over 5 years: results from the breakthrough generations study.
Reproducibility of estradiol and testosterone levels in postmenopausal women over 5 years: results from the breakthrough generations study.
American journal of epidemiology (2014-04-02)
Michael E Jones, Minouk J Schoemaker, Megan Rae, Elizabeth J Folkerd, Mitch Dowsett, Alan Ashworth, Anthony J Swerdlow
ZUSAMMENFASSUNG
Prospective cohort studies examining sex hormones in relation to cancer risk have generally collected blood samples at 1 time point, with an assumption that hormone levels measured in these samples will be reliable markers of true levels at other times. In postmenopausal women, body fat is a major source of estradiol; therefore, changes in adiposity may affect the correlation of single measurements to more relevant long-term averages. To estimate the intraclass correlation coefficient (ICC) for estradiol and testosterone, we collected repeat blood samples from 119 postmenopausal women (average age = 59.4 (standard deviation, 4.7) years) from the United Kingdom during 2004-2005 and again during 2010-2011. The ICCs (adjusted for assay variation) were 0.73 (95% confidence interval: 0.63, 0.82) for total estradiol and 0.59 (95% confidence interval: 0.47, 0.72) for total testosterone. The ICCs were 3%-5% larger after adjustment for change in body mass index (weight (kg)/height (m)(2)) or leptin, which are 2 markers of change in adiposity. There was no increase in ICCs after adjustment for change in age, alcohol consumption, smoking, exercise, time between waking and blood collection, or season. The results suggest that other factors account for within-woman variation in these sex hormones.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Testosteron, purum, ≥99.0% (HPLC)
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17β-Estradiol -Lösung, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
β-Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Estradiol, analytical standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Testosteron, VETRANAL®, analytical standard
Testosteron, European Pharmacopoeia (EP) Reference Standard
Estradiol Hemihydrat, European Pharmacopoeia (EP) Reference Standard