Merck
  • Uptake and efflux of rhenium in cells exposed to rhenium diseleno-ether and tissue distribution of rhenium and selenium after rhenium diseleno-ether treatment in mice.
Uptake and efflux of rhenium in cells exposed to rhenium diseleno-ether and tissue distribution of rhenium and selenium after rhenium diseleno-ether treatment in mice.
Anticancer research (2014-04-03)
Philippe Collery, Gérard Bastian, François Santoni, Ahmed Mohsen, Ming Wei, Thomas Collery, Alain Tomas, Didier Desmaele, Jean D'Angelo
ZUSAMMENFASSUNG
We proposed a new water-soluble rhenium diseleno-ether compound (with one atom of Re and two atoms of Se) and investigated the uptake of Re into the nucleus of malignant cells in culture exposed to the compound for 48 h and its efflux from the nucleus after a post-exposure period of 48 h, as DNA is the main target of Re. We also studied the distribution of both Re and Se in the main organs after an oral administration of 10 or 40 mg/kg Re diseleno-ether to mice for four weeks, five days-a-week. Re and Se concentrations were assayed by inductively coupled plasma mass spectrometry (ICP-MS). Statistical analysis was performed using the Wilcoxon signed-rank test, comparing two related groups. We observed that Re was well incorporated into the nucleus of malignant cells in the most sensitive cells MCF-7, derived from human breast cancer, and that there was no efflux of Re. In contrast, in MCF-7 resistant cells (MCF-7 Mdr and MCF-7 R), A549 and HeLa cells, there was significant efflux of Re from the nucleus after the wash-out period. In mice, an important and dose-dependent uptake of both Re and Se was observed in the liver, with lower concentrations in kidneys. The lowest concentrations were observed in blood, lung, spleen and bones. There was a significant increase of Re concentrations in the blood, liver and kidney in mice treated with Re diseleno-ether at the dose of 40 mg/kg/24 h versus those treated at the dose of 10 mg/kg/24 h. There was a significant increase of Se concentrations in all tissues with the dose of Re diseleno-ether of 10 mg/kg/24 h versus controls, and a significant increase in the liver in mice treated with dose of Re diseleno-ether of 40 mg/kg/24h versus those treated with 10 mg/kg/24 h. We are the first to demonstrate that a compound combining Re and Se in a single molecule, is able to deliver Re and Se to the organism via an oral route, for cancer treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Diethylether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethylether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Diethylether, analytical standard
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Selen, pellets, < 5mm, ≥99.999%
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.076mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Rhenium, wire reel, 0.05m, diameter 0.5mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.15mm, annealed, 99.97%
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%