Merck
  • Efficacy of a novel topical combination of fipronil, (S)-methoprene, eprinomectin and praziquantel against feline urinary bladder worm (Capillaria plica) infection.
Efficacy of a novel topical combination of fipronil, (S)-methoprene, eprinomectin and praziquantel against feline urinary bladder worm (Capillaria plica) infection.
Veterinary parasitology (2014-04-08)
Martin Knaus, Enstela Shukullari, Joseph Rosentel, Steffen Rehbein
ZUSAMMENFASSUNG
Infection with urinary capillarid bladder worms has been observed in cats worldwide. Although considered as generally causing no or little harm, infection with urinary capillarids may be associated with clinical disease which requires an appropriate treatment including the use of anthelmintics. Therefore, the efficacy of a novel topical combination formulation of fipronil 8.3% (w/v), (S)-methoprene 10% (w/v), eprinomectin 0.4% (w/v), and praziquantel 8.3% (w/v) (BROADLINE(®), Merial) was evaluated against urinary capillarids in naturally infected cats. Sixteen European Short Hair cats (5 male, 11 female) with capillarid eggs in their urine pre-treatment were included in the study. At the time of treatment, the cats were approximately ten months to eight years old and weighed 1.6-3.6 kg. Cats were ranked based on decreasing bodyweight and then randomly allocated within replicates of two animals to one of the treatment groups. Each cat in the treated group received one topical application of the combination product at the minimum therapeutic dose of 0.12 mL/kg body weight delivering 10mg fipronil+12 mg (S)-methoprene+0.5mg eprinomectin+10mg praziquantel per kilogram of body weight while the cats allocated to the control group remained untreated. For parasite recovery, identification and count, cats were euthanized humanely 14 days after treatment. All untreated cats harboured Capillaria plica in their urinary bladders (range 4-12), while no capillarids were recovered from the eight treated cats. Thus, the efficacy of the novel topical combination against C. plica was 100%. All cats accepted the treatment well based on post-treatment observations and daily observations thereafter. No adverse events or other health problems were observed during the study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ivermectin
Supelco
Fipronil, PESTANAL®, analytical standard
Supelco
Praziquantel, Pharmaceutical Secondary Standard; Certified Reference Material
Ivermectin, European Pharmacopoeia (EP) Reference Standard
Supelco
Methopren, PESTANAL®, analytical standard
Supelco
Fipronil, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Praziquantel, VETRANAL®, analytical standard
Supelco
Eprinomectin, PESTANAL®, analytical standard, mixture of isomers
Praziquantel, European Pharmacopoeia (EP) Reference Standard
Praziquantel für die Systemeignung, European Pharmacopoeia (EP) Reference Standard