Merck
Excitatory GABAA receptor in autonomic pelvic ganglion neurons innervating bladder.
Biochemical and biophysical research communications (2014-04-08)
Na-Hyun Kim, Seung-Kuy Cha, In Deok Kong
ZUSAMMENFASSUNG
Major pelvic ganglia (MPG) are relay centers for autonomic reflexes such as micturition and penile erection. MPG innervate the urogenital system, including bladder. γ-Aminobutyric acid (GABA) is the major inhibitory neurotransmitter in the mammalian central nervous system, and may also play an important role in some peripheral autonomic ganglia, including MPG. However, the electrophysiological properties and function of GABAA receptor in MPG neurons innervating bladder remain unknown. This study examined the electrophysiological properties and functional roles of GABAA receptors in bladder-innervating neurons identified by retrograde Dil tracing. Neurons innervating bladder showed previously established parasympathetic properties, including small membrane capacitance, lack of T-type Ca(2+) channel expression, and tyrosine-hydroxylase immunoreactivity. GABAA receptors were functionally expressed in bladder innervating neurons, but GABAC receptors were not. GABA elicited strong depolarization followed by increase of intracellular Ca(2+) in neurons innervating bladder, supporting the hypothesis GABA may play an important role in bladder function. These results provide useful information about the autonomic function of bladder in physiological and pathological conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Aminobutansäure, ≥99%
Supelco
4-Aminobutansäure, analytical standard
Sigma-Aldrich
4-Aminobutansäure, BioXtra, ≥99%
Vigabatrin Unreinheit D, European Pharmacopoeia (EP) Reference Standard