Merck
  • ArfGAP3 is a component of the photoreceptor synaptic ribbon complex and forms an NAD(H)-regulated, redox-sensitive complex with RIBEYE that is important for endocytosis.
ArfGAP3 is a component of the photoreceptor synaptic ribbon complex and forms an NAD(H)-regulated, redox-sensitive complex with RIBEYE that is important for endocytosis.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-04-11)
Mayur Dembla, Silke Wahl, Rashmi Katiyar, Frank Schmitz
ZUSAMMENFASSUNG
Ribbon synapses are tonically active synapses in the retina and inner ear with intense vesicle traffic. How this traffic is organized and regulated is still unknown. Synaptic ribbons, large presynaptic structures associated with numerous synaptic vesicles, appear to be essential for this process. The base of the synaptic ribbon is anchored at the active zone and is a hotspot of exocytosis. The synaptic ribbon complex is also important for vesicle replenishment. RIBEYE is a unique and major component of synaptic ribbons. It consists of a unique A-domain and an NAD(H)-binding, C-terminal B-domain. In the present study, we show that the Arf-GTPase activating protein-3 (ArfGAP3), a well characterized regulator of vesicle formation at the Golgi apparatus, is also a component of the synaptic ribbon complex in photoreceptor synapses of the mouse retina and interacts with RIBEYE as shown by multiple, independent approaches. ArfGAP3 binds to RIBEYE(B)-domain in an NAD(H)-dependent manner. The interaction is redox sensitive because NADH is more efficient than the oxidized NAD(+) in promoting ArfGAP3-RIBEYE interaction. RIBEYE competes with the GTP-binding protein Arf1 for binding to ArfGAP3. Thus, binding of RIBEYE(B) to ArfGAP3 could prevent inactivation of Arf1 by ArfGAP3 and provides the synaptic ribbon with the possibility to control Arf1 function. The interaction is relevant for endocytic vesicle trafficking because overexpression of ArfGAP3 in photoreceptors strongly inhibited endocytotic uptake of FM1-43.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, Grade AA-1
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 20 mg (per vial)