Merck
Developmental toxicity in rare minnow (Gobiocypris rarus) embryos exposed to Cu, Zn and Cd.
Ecotoxicology and environmental safety (2014-04-15)
Bin Zhu, Lei Liu, Dong-Liang Li, Fei Ling, Gao-Xue Wang
ZUSAMMENFASSUNG
Using rare minnow (Gobiocypris rarus) embryos as experimental model, the developmental toxicity of Cu, Zn and Cd were investigated following exposure to 0.001-1.000mg/L for 72h, and the toxicity effects were evaluated by larval malformation rate, heart rate, pericardial area, spontaneous movements, tail length, enzyme activities and biomarker genes. Our results revealed that increased malformation rate provide a gradual dose-response relationship, and the most pronounced morphological alteration was heart and body malformations. Values of 72h EC50 with their 95 percent confidence intervals on G. rarus embryos were 0.103 (0.072-0.149)mg/L for Cu, 0.531 (0.330-1.060)mg/L for Zn, 0.219 (0.147-0.351)mg/L for Cd. Enzyme activities can be regard as a type of low-dose stimulation and high-dose inhibition. Stress and metabolism-related genes (hsp70, cyp1a and mt) were significantly up-regulated, development-related genes (wnt8a, vezf1 and mstn) were significantly down-regulated after the treatment by Cu, Zn and Cd. Overall, the present study points out Cu, Zn and Cd are highly toxic to G. rarus embryos. The information presented in this study will be helpful in fully understanding the toxicity induced by Cu, Zn and Cd in fish embryos.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zink, dust, <10 μm, ≥98%
Sigma-Aldrich
Zink, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zink, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Kupfer, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, powder, 99.99% trace metals basis
Sigma-Aldrich
Zink, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zink, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zink, purum, powder
Sigma-Aldrich
Kupfer, powder, <75 μm, 99%
Sigma-Aldrich
Kupfer, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Kupfer, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Kupfer, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Zink, granular, 30-100 mesh, 99%
Sigma-Aldrich
Kupfer, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Zink, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Kupfer, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Kupfer, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Zink, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95+%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Kupfer, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Zink, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Kupfer, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Kupfer, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Zink, mossy, ≥99%
Kupfer, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh